Tisk…

2,4-Dimethyl-6-iodoaniline ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H51875.03 H51875.06
H51875.03EA 2720 CZK
H51875.03 H51875.06
2,4-Dimethyl-6-iodoaniline ≥98%
2,4-Dimethyl-6-iodoaniline

Purity: 98% CAS No.: 4102-54-9 Molecular Formula: C8H10IN Formula Weight: 247.08 Melting Point: 64-65° UN No.: 2811 MDL No.: MFCD07779005

Vzorec: C₈H₁₀IN
Bod tání: 64…65 °C
Storage Temperature: Ambient
MDL Number: MFCD07779005
CAS číslo: 4102-54-9
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR