Tisk…

2-Chloro-3-(trifluoromethyl)quinoxaline 96%

Dodavatel: Thermo Fisher Scientific
Varování

H50549.03 H50549.06
H50549.03EA 2360 CZK
H50549.03 H50549.06
2-Chloro-3-(trifluoromethyl)quinoxaline 96%
2-Chloro-3-(trifluoromethyl)quinoxaline

Purity: 96% CAS No.: 254732-51-9 Molecular Formula: C9H4ClF3N2 Formula Weight: 232.59 MDL No.: MFCD00243891

Vzorec: C₉H₄ClF₃N₂
Storage Temperature: Ambient
MDL Number: MFCD00243891
CAS číslo: 254732-51-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR