Tisk…

Ethyl-6-aminohexanoate 98%

Dodavatel: Thermo Fisher Scientific

H50296.03 H50296.MD
H50296.03EA 9920 CZK
H50296.03 H50296.MD
Ethyl-6-aminohexanoate 98%
Ethyl-6-aminohexanoate

Purity: 98% CAS No.: 371-34-6 Molecular Formula: C8H17NO2 Formula Weight: 159.23 MDL No.: MFCD00233495

Vzorec: C₈H₁₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00233495
CAS číslo: 371-34-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR