Tisk…

2-(Cyclopentyloxy)-3-pyridineboronic acid pinacol ester 96%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-Cyclopentyloxypyridine-3-boronic acid pinacol ester

H50110.03
H50110.03EA 3350 CZK
H50110.03
2-(Cyclopentyloxy)-3-pyridineboronic acid pinacol ester 96%
2-(Cyclopentyloxy)-3-pyridineboronic acid pinacol ester

Purity: 96% CAS No.: 1073371-90-0 Molecular Formula: C16H24BNO3 Formula Weight: 289.19 MDL No.: MFCD07781170

Vzorec: C₁₆H₂₄BNO₃
MW: 289.18
Storage Temperature: Ambient
MDL Number: MFCD07781170
CAS číslo: 1073371-90-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR