Tisk…

6-Bromoimidazo[1,2-a]pyridine ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H34453.03 H34453.06
H34453.03EA 1130 CZK
H34453.03 H34453.06
6-Bromoimidazo[1,2-a]pyridine ≥98%
6-Bromoimidazo[1,2-a]pyridine

Purity: 98% CAS No.: 6188-23-4 Molecular Formula: C7H5BrN2 Formula Weight: 197.03 Melting Point: 78-80° MDL No.: MFCD06496200

Vzorec: C₇H₅BrN₂
Bod tání: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD06496200
CAS číslo: 6188-23-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR