Tisk…

3-(4-Chloro-3-fluorophenyl)propionic acid 96%

Dodavatel: Thermo Fisher Scientific
Varování

H34205.03 H34205.06
H34205.03EA 1250 CZK
H34205.03 H34205.06
3-(4-Chloro-3-fluorophenyl)propionic acid 96%
3-(4-Chloro-3-fluorophenyl)propionic acid

Purity: 96% CAS No.: 881189-65-7 Molecular Formula: C9H8ClFO2 Formula Weight: 202.61 Melting Point: 80-84° MDL No.: MFCD04116058

Vzorec: C₉H₈ClFO₂
Bod tání: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD04116058
CAS číslo: 881189-65-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR