Tisk…

2-Hydroxy-3-(trifluoromethyl)benzaldehyde ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

H32858.MD H32858.03
H32858.MDEA 2840 CZK
H32858.MD H32858.03
2-Hydroxy-3-(trifluoromethyl)benzaldehyde ≥98%
2-Hydroxy-3-(trifluoromethyl)benzaldehyde

Purity: 98+% CAS No.: 336628-67-2 Molecular Formula: C8H5F3O2 Formula Weight: 190.12 Melting Point: 54-59° MDL No.: MFCD03788529

Vzorec: C₈H₅F₃O₂
Bod tání: 54…59 °C
Storage Temperature: Ambient
MDL Number: MFCD03788529
CAS číslo: 336628-67-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR