Tisk…

5-Fluoro-2-nitrobenzonitrile ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H32756.03 H32756.06
H32756.03EA 1810 CZK
H32756.03 H32756.06
5-Fluoro-2-nitrobenzonitrile ≥97%
5-Fluoro-2-nitrobenzonitrile

Purity: 97% CAS No.: 50594-78-0 Molecular Formula: C7H3FN2O2 Formula Weight: 166.11 Melting Point: 101-105° UN No.: 3439 MDL No.: MFCD01632196

Vzorec: C₇H₃FN₂O₂
MW: 166.11
Bod tání: 101…105 °C
Storage Temperature: Ambient
MDL Number: MFCD01632196
CAS číslo: 50594-78-0
UN: 3439
ADR: 6.1 III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR