Tisk…

4,5-Dimethyl-o-phenylenediamine, (∼15% H₂O) 98%

Dodavatel: Thermo Fisher Scientific
Varování

H32695.06 H32695.14
H32695.06EA 2190 CZK
H32695.06 H32695.14
4,5-Dimethyl-o-phenylenediamine, (∼15% H₂O) 98%
4,5-Dimethyl-o-phenylenediamine

Purity: 98% CAS No.: 3171-45-7 Molecular Formula: C8H12N2 Formula Weight: 136.20 Storage Temperature: 2°C to 8°C Melting Point: 127-129° UN No.: UN2811 MDL No.: MFCD00007729

Vzorec: C₈H₁₂N₂
Bod tání: 127…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007729
CAS číslo: 3171-45-7
EINECS: 221-635-1
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR