Tisk…

3-Bromo-1-propylboronic acid pinacol ester ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

H30646.03 H30646.06 H30646.14
H30646.03EA 1720 CZK
H30646.03 H30646.06 H30646.14
3-Bromo-1-propylboronic acid pinacol ester ≥98%
3-Bromo-1-propylboronic acid pinacol ester

Purity: 98% CAS No.: 124215-44-7 Molecular Formula: C9H18BBrO2 Formula Weight: 248.96 Boiling Point: 61-64°/0.35mm MDL No.: MFCD10567053

Vzorec: C₉H₁₈BBrO₂
Bod varu: 61…64 °C (0,35 mmHg)
Hustota: 1,177
Storage Temperature: Ambient
MDL Number: MFCD10567053
CAS číslo: 124215-44-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR