Tisk…

(±)-3-Hydroxybutyric acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

H30426.06 H30426.14
H30426.06EA 3100 CZK
H30426.06 H30426.14
(±)-3-Hydroxybutyric acid ≥98%
(±)-3-Hydroxybutyric acid

Purity: 98% CAS No.: 300-85-6 Molecular Formula: C4H8O3 Formula Weight: 104.10 Odour: Odourless Melting Point: 36-39° MDL No.: MFCD00004546

Vzorec: C₄H₈O₃
Bod varu: 269 °C (1013 hPa)
Bod tání: 36…39 °C
Hustota: 1,26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004546
CAS číslo: 300-85-6
EINECS: 206-099-9
Merck Index: 14,4816

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR