Tisk…

1,7-Dihydroxynaphthalene ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Naphthalene-1,7-diol

H28261.14 H28261.06
H28261.14EA 6080 CZK
H28261.14 H28261.06
1,7-Dihydroxynaphthalene ≥97%
1,7-Dihydroxynaphthalene

Purity: 97% CAS No.: 575-38-2 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 180-184° MDL No.: MFCD00035720

Vzorec: C₁₀H₈O₂
Bod varu: 375 °C (1013 hPa)
Bod tání: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00035720
CAS číslo: 575-38-2
EINECS: 209-383-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR