Tisk…

4-(4-Dimethylaminophenylazo)benzoic acid sodium salt Indicator

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: p-Methyl red, Sodium 4-(4-dimethylaminophenylazo)benzoate

H27830.06
H27830.06EA 6390 CZK
H27830.06
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt Indicator
4-(4-Dimethylaminophenylazo)benzoic acidsodium salt

CAS No.: 845-46-5 Molecular Formula: C15H14N3NaO2 Formula Weight: 291.28 Melting Point: 300° MDL No.: MFCD00020350

Vzorec: C₁₅H₁₄N₃O₂Na
MW: 291.28 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00020350
CAS číslo: 845-46-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR