Tisk…

7-Aminoenanthic acid 98%

Dodavatel: Thermo Fisher Scientific

H27519.03 H27519.06
H27519.03EA 4090 CZK
H27519.03 H27519.06
7-Aminoenanthic acid 98%
7-Aminoenanthic acid

Purity: 98% CAS No.: 929-17-9 Molecular Formula: H2N(CH2)6CO2H Formula Weight: 145.19 Melting Point: 194-196° Beilstein Reference: 906887

Vzorec: C₇H₁₅NO₂
Bod tání: 194…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00008242
CAS číslo: 929-17-9
EINECS: 213-197-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR