Tisk…

1-Butyl-4-methylpyridiniumtetrafluoroborate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

H27100.06 H27100.18
H27100.06EA 2660 CZK
H27100.06 H27100.18
1-Butyl-4-methylpyridiniumtetrafluoroborate ≥99%
1-Butyl-4-methylpyridiniumtetrafluoroborate

Purity: 99% CAS No.: 343952-33-0 Molecular Formula: C10H16BF4N Formula Weight: 237.06 Refractive Index: 1.4

Vzorec: C₁₀H₁₆BF₄N
Hustota: 1,2
Storage Temperature: Ambient
MDL Number: MFCD03095460
CAS číslo: 343952-33-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR