Tisk…

1-Butylpyridinium bromide ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: N-butylpyridinium bromide, 1-n-Butylpyridinium bromide

H26995.18 H26995.06
H26995.18EA 6040 CZK
H26995.18 H26995.06
1-Butylpyridinium bromide ≥99%
1-Butylpyridinium bromide

Purity: 99% CAS No.: 874-80-6 Molecular Formula: C9H14BrN Formula Weight: 216.12 Melting Point: 105-108° MDL No.: MFCD00671496

Vzorec: C₉H₁₄BrN
MW: 216.12
Bod tání: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00671496
CAS číslo: 874-80-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR