Tisk…

4-(Methylthio)phenylacetic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H26888.03 H26888.06
H26888.03EA 862 CZK
H26888.03 H26888.06
4-(Methylthio)phenylacetic acid ≥97%
4-(Methylthio)phenylacetic acid

Purity: 97% CAS No.: 16188-55-9 Molecular Formula: C9H10O2S Formula Weight: 182.24 Melting Point: 98-102° MDL No.: MFCD00192325

Vzorec: C₉H₁₀O₂S
MW: 182.24
Bod tání: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00192325
CAS číslo: 16188-55-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR