Tisk…

3-Bromo-5-(trifluoromethyl)benzaldehyde ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H26197.03 H26197.06
H26197.03EA 982 CZK
H26197.03 H26197.06
3-Bromo-5-(trifluoromethyl)benzaldehyde ≥97%
3-Bromo-5-(trifluoromethyl)benzaldehyde

Purity: 97% CAS No.: 477535-41-4 Molecular Formula: C8H4BrF3O Formula Weight: 253.02 Refractive Index: 1.5110

Vzorec: C₈H₄BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD08059506
CAS číslo: 477535-41-4
EINECS: None

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR