Tisk…

3,6-Difluorophthalic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

H26096.03
H26096.03EA 5670 CZK
H26096.03
3,6-Difluorophthalic acid ≥98%
3,6-Difluorophthalic acid

Purity: 98% CAS No.: 651-97-8 Molecular Formula: C8H4F2O4 Formula Weight: 202.11 Melting Point: ca 184° dec. MDL No.: MFCD03840499

Vzorec: C₈H₄F₂O₄
Storage Temperature: Ambient
MDL Number: MFCD03840499
CAS číslo: 651-97-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR