Tisk…

N,N-Dimethyl-1-dodecylamine N-oxide 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H26065.14 H26065.06
H26065.14EA 18330 CZK
H26065.14 H26065.06
N,N-Dimethyl-1-dodecylamine N-oxide 95%
N,N-Dimethyl-1-dodecylamine N-oxide

Purity: 95% CAS No.: 1643-20-5 Molecular Formula: C14H31NO Formula Weight: 229.40 Melting Point: 120-123° Beilstein Reference: 1769927 UN No.: 3259 MDL No.: MFCD00002049

Vzorec: C₁₄H₃₁NO
Bod tání: 120…123 °C
Hustota: 0,996
Storage Temperature: Refrigerator
MDL Number: MFCD00002049
CAS číslo: 1643-20-5
EINECS: 216-700-6
UN: 3259
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR