Tisk…

3-(3,4-Dimethoxybenzoyl)propionic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

H25806.14 H25806.06
H25806.14EA 10510 CZK
H25806.14 H25806.06
3-(3,4-Dimethoxybenzoyl)propionic acid ≥98%
3-(3,4-Dimethoxybenzoyl)propionic acid

Purity: 98% CAS No.: 5333-34-6 Molecular Formula: C12H14O5 Formula Weight: 238.24 Melting Point: 159-161° MDL No.: MFCD00033286

Vzorec: C₁₂H₁₄O₅
Bod tání: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00033286
CAS číslo: 5333-34-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR