Tisk…
pH/mV/°C meter, handheld, GMH 3511
Dodavatel: Greisinger
604953 605021
GRSG604953EA 0 CZK
GRSG604953 GRSG605021
pH/mV/°C meter, handheld, GMH 3511
Meřiče elektrochemické Meřiče pH a multimetry
For comfortable measurements of pH value and temperature.
  • ORP mode allows for automatic conversion to hydrogen system electrodes
  • Temperature compensation
  • Automatic buffer detection
  • Rating function of electrode‘s quality
  • Analog output
Easier operation ensured by a menu reduced to 5 points. Minimal measuring effort because of maintenance-free gel-electrode and automatic temperature compensation.
Informace o dodávce: Model GMH 3511: Meter only; electrodes and accessories sold separately. Supplied with device, battery, calibration protocol, and manual.
More Product Information