Tisk…
Adhesive seal, SILVERseal™
Dodavatel: Greiner Bio-One
676090_1200
GREI676090_1200EA 39510 CZK
GREI676090_1200
Adhesive seal, SILVERseal™
Těsnění Těsnění mikrodestičky
SILVERseal™ is a heat resistant, cold-resistant and pierceable aluminium foil, which is suitable for both PCR applications and sample storage.
  • Temperature resistant from −80 to 110 °C
  • Coated with an acrylate adhesive, which remains flat when removed from its paper backing, and can be easily applied to all microplate types
  • A double perforation makes it possible to tear off the projecting adhesive sealer and thus improves the stacking of sealed microplates
VIEWseal™ is an adhesive sealing film coated with a silicone adhesive, which only sticks when theMore Product Information