Tisk…
Coagulation tubes, VACUETTE®
Dodavatel: Greiner Bio-One
454321_1200 454327_1200 455322_1200 454322_1200 454325_1200 474321_1200 454332_1200 454387_1200 474327 454334_1200
GREI454321_1200EA 5460 CZK
GREI454321_1200 GREI454327_1200 GREI455322_1200 GREI454322_1200 GREI454325_1200 GREI474321_1200 GREI454332_1200 GREI454387_1200 GREI474327 GREI454334_1200
Coagulation tubes, VACUETTE®
Systémy pro klinickou diagnostiku Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
VACUETTE® tubes contain sodium citrate solutions in concentrations of 3,2% or 3,8% and are used for assays in haemostaseology. The mixing of 9 parts blood to 1 part sodium citrate takes place immediately after the blood is drawn by inverting the tube, causing anticoagulation through the binding of calcium.
  • Clear as glass but virtually unbreakable
  • Screw caps for increased safety and simplified opening
  • Colour coded rings offer additional visual identification of tube characteristics
  • Longer shelf-life due to tube design
  • Compatible with all common analyser
More Product Information