Tisk…
Blood collection tubes with NH trace elements sodium heparin, VACUETTE®
Dodavatel: Greiner Bio-One
456080_1200
GREI456080_1200EA 6210 CZK
GREI456080_1200
Blood collection tubes with NH trace elements sodium heparin, VACUETTE®
Systémy pro klinickou diagnostiku Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Tubes are designed to test trace elements such as aluminium, lead, cadmium, chrome, iron, fluoride, cobalt, copper, lithium, manganese, molybdenum, mercury, selenium, or thallium etc.
  • Sterile
These tubes are supplied with premium safety twist cap with black ring at the cap.