Tisk…
Racks and Boxes for Microcentrifuge Tubes
Dodavatel: GLW
GSTY G50B G50G G50R VST
391-8855EA 50.2 CZK
391-8855 391-8856 391-8857 391-8858 GLWAVST
Racks and Boxes for Microcentrifuge Tubes
Krabice Krabičky na zkumavky
Styro racks are made of heavy expanded polystyrene which are used in freezer down to -90 °C. System box is made of cardboard and is designed to securely enclose two styro rack boxes.
  • Not autoclavable
  • For 1,5 and 2,0 ml tubes up to 12 mm diameter