Tisk…
PLC purification systems
Dodavatel: GILSON LCMS
21140001 21140004 21140002 21140005
GILI21140001EA 1211460 CZK
GILI21140001 GILI21140004 GILI21140002 GILI21140005
PLC purification systems
Systémy chromatografické HPLC systémy
PLC 2050/2250/2500 purification systems are designed for preparative liquid chromatography (PLC) to aspirate mobile phase solvents, form gradients, inject liquid samples, and provide solutions for a preparative column for high performance liquid chromatography (HPLC), centrifugal partition chromatography (CPC), or counter-current chromatography (CCC) applications.
  • PLC 2050: Flow rates up to 50 ml/min with pressure up to 300 bar
  • PLC 2250: Flow rates up to 250 ml/min with pressure up to 230 bar
  • PLC 2500: Flow rates up to 500 ml/min with pressure up to 110
More Product Information