Tisk…
Ergonomic washable silicone mouse, GETT GCQ®
Dodavatel: GETT GERATETECHNIK
KH24207
GETTKH24207EA 2330 CZK
GETTKH24207
Ergonomic washable silicone mouse, GETT GCQ®
Počítačová myš a podložka pod myš
This silicone mouse is a true all-rounder. It can be operated ergonomically and, thanks to the silicone surface, is perfect for application areas with high hygiene requirements.
  • Medical grade IP 68 mouse is perfect for applications with high hygiene requirements
  • Fully washable and disinfectable
  • Silicone cap to protect the USB connector from splashing water during cleaning
  • Ergonomic functionality with two mouse buttons as well as 3 scroll buttons
  • Laser sensor
  • Resolution: 1000 dpi
  • Button lifetime: >=2 mio. actuations
  • Engineered in Germany
  • 2-year warranty
More Product Information