Tisk…
Advanced MB, InduProof®
Dodavatel: GETT GERATETECHNIK
KG17203 KG17204
GETTKG17203EA 9320 CZK
GETTKG17203 GETTKG17204
Advanced MB, InduProof®
Klávesnice
Rugged, water- and dust-proof silicone keyboard with integrated mouse button.
  • Rugged, durable keyboard with 104 keys and integrated mouse button
  • Comfortable & ergonomic operation also with protective gloves made of silicone or latex
  • IP 68 protection, can be submerged into liquids
  • Easy cleaning and disinfection - even under tap water or by immersion
  • Increased chemical and mechanical resistance due to special surface treatment
  • VESA-M5-boreholes (distance 75×75 mm)
  • 4 years warranty on technical function
  • Made in Germany
The InduProof® Advanced has aMore Product Information