Tisk…

Písek, Crystalline 0,1-0,5 mm, washed, calcinated

Dodavatel: GERBER FUNKE

Synonyms: Silicium dioxide, Silicium (IV) oxide, Silicon dioxide

5460 5461 5462 5464
GERB5460EA 346 CZK
GERB5460 GERB5461 GERB5462 GERB5464
Písek, Crystalline 0,1-0,5 mm, washed, calcinated
Písek

Crystalline, calcined quality.

Vzorec: SiO₂
MW: 60.08
Bod tání: 1700
Hustota: 2.6
MDL Number: MFCD02100519
CAS číslo: 7631-86-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR