Tisk…

Anti-SARS-CoV-2 Spike Protein (RBD) Human Monoclonal Antibody [clone: HC2001]

Dodavatel: Genscript
A02038-100 A02038-1
GENSA02038-100EA 17800 CZK
GENSA02038-100 GENSA02038-1
Anti-SARS-CoV-2 Spike Protein (RBD) Human Monoclonal Antibody [clone: HC2001]
Protilátky
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease 2019 (COVID-19). SARS-CoV-2 contains glycosylated spike (S) protein, which is composed of S1 subunit and S2 subunit. The S1 contains a receptor-binding domain (RBD) that can bind to ACE2 receptor on target cells.
SARS-CoV-2 Spike S1 Antibody (HC2001), Human Chimeric is produced from cell culture in vitro under conditions free from animal-derived components.

Type: Primary
Antigen: Protein
Clonality: Monoclonal
Clone: HC2001 (More Product Information
Objednat

Produktová specifikace

Environmentally Preferable
Název antigenu SARS-CoV-2 Spike Protein (RBD)
Synonyma antigenu Receptor-binding domain/RBD
Druh protilátky Primární
Klonovalita Monoclonal
Spojení Unconjugated
Klon HC2001
Reaktivita Human
Hostitel Human
Identifikační číslo genu MN908947
Izotyp IgG1
ELISA Yes
Fragment protilátky Whole Molecule
Modifikace Humanization
Epitop Confidentiality
Imunogen Confidentiality
Počet aminokyselin Confidentiality
Sekvence Confidentiality
Formát Purified antibody
Formulář liquid
Molekulární hmotnost 150 kDa
Čištění Protein A
Koncentrace 1 mg/ml, supplied in PBS, pH 7.4, containing 0.02% sodium azide.
Křížová adsorbce No
Skladovací pufr PBS, pH 7.4, containing 0.02% sodium azide.
Skladovací teplota ­20° C
Transportní teplota 4° C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR