Tisk…

Anti-ACTB Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Genscript
A00730-200
GENSA00730-200EA 17510 CZK
GENSA00730-200
Anti-ACTB Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Tag
Antigen: ACTB (actin, beta)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR