Tisk…

Anti-DYKDDDDK Tag Mouse Monoclonal Antibody

Dodavatel: Genscript
A00187-100
GENSA00187-100EA 6980 CZK
GENSA00187-100
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody
Protilátky
Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR