Tisk…
spin columns, C18
Dodavatel: G-Biosciences
786-930 786-931
GENO786-930EA 7390 CZK
GENO786-930 GENO786-931
spin columns, C18
Filtry Filtry centrifugační
These C18 spin columns are ready-to-use micro centrifuge columns for peptide clean up and concentration. The columns consist of porous C18 reverse-phase resin that has a particle size of ~15 µm and a pore size of 300 Å. The resin offers highly efficient binding and recovery of peptides and is ideal for mass spectrometry and other peptide related applications.
  • Rapid peptide purification
  • Yields high quality spectra with significant noise reduction
  • Easy to handle, no specialised equipment required
  • End capped to block reactive surface sianol groups
  • 10 mg C18
More Product Information