Tisk…
Avantor® ACE® Generix® Phenyl-Hexyl & Generix® SIL, Analytical HPLC Columns, 3 µm
Dodavatel: Avantor
GEN3SIL0502/3EA 0 CZK
GEN3SIL0502/3 GEN3SIL1046/3 GEN3SIL1546/3 GEN3PHEX0502/3 GEN3PHEX1046/3 GEN3PHEX1546/3 GEN-SIL-GD2 GEN-SIL-GD GEN-PHEX-GD2 GEN-PHEX-GD GEN-3SIL-0502 GEN-3SIL-7502 GEN-3SIL-1002 GEN-3SIL-1202 GEN-3SIL-1502 GEN-3SIL-0503 GEN-3SIL-7504 GEN-3SIL-1004 GEN-3SIL-1204 GEN-3SIL-1504 GEN-3SIL-0546 GEN-3SIL-7546 GEN-3SIL-1046 GEN-3SIL-1246 GEN-3SIL-1546 GEN-3SIL-7503 GEN-3SIL-1003 GEN-3SIL-1203 GEN-3SIL-1503 GEN-3SIL-0504
Avantor® ACE® Generix® Phenyl-Hexyl & Generix® SIL, Analytical HPLC Columns, 3 µm
Kolony chromatografické
These high performance HPLC columns contain high purity spherical silica.
  • Ultra-narrow particle size distribution for excellent column efficiency
  • Sharp, highly symmetrical peaks for acidic, basic and neutral analytes
  • Excellent column to column and batch to batch reproducibility
Each column is individually tested and supplied with a test chromatogram.