Tisk…
Cryogenic Tapes for Laboratory Use
Dodavatel: GA
TJTA-13C1-50WH TJTA-13C1-50RE TJTA-13C1-50YE TJTA-13C1-50BL TJTA-13C1-50OR TJTA-13C1-50GA TJTA13C3-100WH TJTA13C3-100RE TJTA13C3-100YE TJTA13C3-100BL TJTA13C3-100OR TJTA13C3-100GA TJTA-19C1-50WH TJTA-19C1-50RE TJTA-19C1-50YE TJTA-19C1-50BL TJTA-19C1-50OR TJTA-19C1-50GA TJTA19C3-100WH TJTA19C3-100RE TJTA19C3-100YE TJTA19C3-100BL TJTA19C3-100OR TJTA19C3-100GA TJTA-25C1-50WH TJTA-25C1-50RE TJTA-25C1-50YE TJTA-25C1-50BL TJTA-25C1-50OR TJTA-25C1-50GA TJTA25C3-100WH TJTA25C3-100RE TJTA25C3-100YE TJTA25C3-100BL TJTA25C3-100OR TJTA25C3-100GA TJTA-70C1-50WH TJTA-70C1-50RE TJTA-70C1-50YE TJTA-70C1-50BL TJTA-70C1-50OR TJTA-70C1-50GA TJTA70C3-100WH TJTA70C3-100RE TJTA70C3-100YE TJTA70C3-100BL TJTA70C3-100OR TJTA70C3-100GA
GAINTJTA-13C1-50WHEA 875 CZK
GAINTJTA-13C1-50WH GAINTJTA-13C1-50RE GAINTJTA-13C1-50YE GAINTJTA-13C1-50BL GAINTJTA-13C1-50OR GAINTJTA-13C1-50GA GAINTJTA13C3-100WH GAINTJTA13C3-100RE GAINTJTA13C3-100YE GAINTJTA13C3-100BL GAINTJTA13C3-100OR GAINTJTA13C3-100GA GAINTJTA-19C1-50WH GAINTJTA-19C1-50RE GAINTJTA-19C1-50YE GAINTJTA-19C1-50BL GAINTJTA-19C1-50OR GAINTJTA-19C1-50GA GAINTJTA19C3-100WH GAINTJTA19C3-100RE GAINTJTA19C3-100YE GAINTJTA19C3-100BL GAINTJTA19C3-100OR GAINTJTA19C3-100GA GAINTJTA-25C1-50WH GAINTJTA-25C1-50RE GAINTJTA-25C1-50YE GAINTJTA-25C1-50BL GAINTJTA-25C1-50OR GAINTJTA-25C1-50GA GAINTJTA25C3-100WH GAINTJTA25C3-100RE GAINTJTA25C3-100YE GAINTJTA25C3-100BL GAINTJTA25C3-100OR GAINTJTA25C3-100GA GAINTJTA-70C1-50WH GAINTJTA-70C1-50RE GAINTJTA-70C1-50YE GAINTJTA-70C1-50BL GAINTJTA-70C1-50OR GAINTJTA-70C1-50GA GAINTJTA70C3-100WH GAINTJTA70C3-100RE GAINTJTA70C3-100YE GAINTJTA70C3-100BL GAINTJTA70C3-100OR GAINTJTA70C3-100GA
Cryogenic Tapes for Laboratory Use
Pásky
Continuous writable tape for cryogenic use. Provided with a strong permanent adhesive, this cryo tape will stay firmly attached during prolonged exposure to low temperature conditions, as well as repeated freeze-thaw cycles. Available in assorted colours.
  • Temperature range: −196 to +110 °C
  • Designed for use in harsh cryogenic conditions
  • Deep-freeze permanent adhesive ensures the tape won't peel or crack when exposed to liquid nitrogen
  • Continuous tape format allows use of as much or little as necessary
Waterproof, resistant to liquid nitrogen, dry ice,More Product Information