Tisk…
Cryogenic Tapes for Frozen Surfaces
Dodavatel: GA
TFS-13C1-50WH TFS-13C1-50YE TFS-13C1-50GA TFS-13C1-50BL TFS-13C1-50OR TFS-13C1-50RE TFS-13C3-100WH TFS-13C3-100YE TFS-13C3-100GA TFS-13C3-100BL TFS-13C3-100OR TFS-13C3-100RE TFS-19C1-50WH TFS-19C1-50YE TFS-19C1-50GA TFS-19C1-50BL TFS-19C1-50OR TFS-19C1-50RE TFS-19C3-100WH TFS-19C3-100YE TFS-19C3-100GA TFS-19C3-100BL TFS-19C3-100OR TFS-19C3-100RE TFS-25C1-50WH TFS-25C1-50YE TFS-25C1-50GA TFS-25C1-50BL TFS-25C1-50OR TFS-25C1-50RE TFS-25C3-100WH TFS-25C3-100YE TFS-25C3-100GA TFS-25C3-100BL TFS-25C3-100OR TFS-25C3-100RE
GAINTFS-13C1-50WHEA 995 CZK
GAINTFS-13C1-50WH GAINTFS-13C1-50YE GAINTFS-13C1-50GA GAINTFS-13C1-50BL GAINTFS-13C1-50OR GAINTFS-13C1-50RE GAINTFS-13C3-100WH GAINTFS-13C3-100YE GAINTFS-13C3-100GA GAINTFS-13C3-100BL GAINTFS-13C3-100OR GAINTFS-13C3-100RE GAINTFS-19C1-50WH GAINTFS-19C1-50YE GAINTFS-19C1-50GA GAINTFS-19C1-50BL GAINTFS-19C1-50OR GAINTFS-19C1-50RE GAINTFS-19C3-100WH GAINTFS-19C3-100YE GAINTFS-19C3-100GA GAINTFS-19C3-100BL GAINTFS-19C3-100OR GAINTFS-19C3-100RE GAINTFS-25C1-50WH GAINTFS-25C1-50YE GAINTFS-25C1-50GA GAINTFS-25C1-50BL GAINTFS-25C1-50OR GAINTFS-25C1-50RE GAINTFS-25C3-100WH GAINTFS-25C3-100YE GAINTFS-25C3-100GA GAINTFS-25C3-100BL GAINTFS-25C3-100OR GAINTFS-25C3-100RE
Cryogenic Tapes for Frozen Surfaces
Pásky
These tapes are intended for identifying frozen containers and surfaces under deep freeze conditions. Ideal for long-term cryo preservation of samples in liquid nitrogen.
  • Temperature range: −196 to +100 °C (−321 to 212 °F)
  • Quickly adheres to already frozen containers and boxes; ideal for long-term storage in cryogenic and deep freeze conditions
  • Continuous format allows use of as much or little as necessary
  • Available in a variety of colours for easy and convenient colour coding
Cryogenic tape for the identification of already frozen containers such asMore Product Information