Tisk…
Transparent Cryogenic Tapes for Frozen Surfaces
Dodavatel: GA
TAQ-13C1-50 TAQ-13C3-100 TAQ-19C1-50 TAQ-19C3-100 TAQ-25C1-50 TAQ-25C3-100 TAQ-38C1-50 TAQ-38C3-100
GAINTAQ-13C1-50EA 955 CZK
GAINTAQ-13C1-50 GAINTAQ-13C3-100 GAINTAQ-19C1-50 GAINTAQ-19C3-100 GAINTAQ-25C1-50 GAINTAQ-25C3-100 GAINTAQ-38C1-50 GAINTAQ-38C3-100
Transparent Cryogenic Tapes for Frozen Surfaces
Pásky
Transparent tape for labelling already frozen containers under deep freeze conditions (–80 °C). This cryogenic tape can be inscribed with permanent cryogenic markers, and is ideal for identifying containers intended for long-term storage in cryogenic (–196 °C) and deep freeze (–80 °C) conditions.
  • Temperature range: –196 to +150 °C (–321 to +302 °F)
  • Tape can adhere to already frozen containers and boxes
  • Ideal for long-term storage in cryogenic (–196 °C) and deep freeze (–80 °C) conditions
  • Strong adhesion to surfaces, even in water bath thawing applications
More Product Information