Tisk…
Printable tags for high-profile PCR tubes and tube strips
Dodavatel: GA
PID-1C1-0.5MC1 PID-1C1-0.5WH PID-1C1-0.5YE PID-1C3-1MC1 PID-1C3-1WH PID-1C3-1YE
GAINPID-1C1-0.5MC1EA 1520 CZK
GAINPID-1C1-0.5MC1 GAINPID-1C1-0.5WH GAINPID-1C1-0.5YE GAINPID-1C3-1MC1 GAINPID-1C3-1WH GAINPID-1C3-1YE
Printable tags for high-profile PCR tubes and tube strips
Štítky
Thermal-transfer printable tags for the identification of high-profile PCR tubes, strips and plates, it allows PCR tubes to be labelled with far more information, in a fraction of the time.
  • Versatile design allows PCR tubes, strips, and plates to be tagged in a variety of configurations and styles
  • Quick, bird’s eye view of all sample information
  • Identify PCR samples with more information than cap dots and tube side rectangular labels combined
  • Thermal-transfer printable, allows printing with barcodes for easy tracking
  • They are safe for use in thermocyclers,
More Product Information