Tisk…
Laboratory warning labels
Dodavatel: GA
L-001-1R L-002-1R L-003-0.5R L-004-1P L-004-1R L-007-0.5R L-008-0.5R L-010-0.5R
GAINL-001-1REA 875 CZK
GAINL-001-1R GAINL-002-1R GAINL-003-0.5R GAINL-004-1P GAINL-004-1R GAINL-007-0.5R GAINL-008-0.5R GAINL-010-0.5R
Laboratory warning labels
Štítky
Preprinted, and blank warning labels for the identification of equipment, containers, and material.
  • Coated with either a permanent or removable adhesive
  • Waterproof, low and high-temperature resistant
  • Can be applied on plastic, glass, cardboard, and metal surfaces
Labels are designed to alert personnel to safety risks in the laboratory environment, and to communicate special use, handling, and disposal. With a wide temperature resistance range, these labels can be used in lab freezers and refrigerators, at room temperature (doors, drawers, walls,More Product Information