Tisk…
Paper labels for thermal-transfer printers
Dodavatel: GA
GPB-122C3-1WH GPB-19C1-3WH GPB28C1-1WH-C6 GPB-28C3-4WH
GAINGPB-122C3-1WHEA 2430 CZK
GAINGPB-122C3-1WH GAINGPB-19C1-3WH GAINGPB28C1-1WH-C6 GAINGPB-28C3-4WH
Paper labels for thermal-transfer printers
Štítky
For general office and laboratory organization, warehouses, and shipping facilities.
  • Strong adhesive ensures labels won't wrinkle or flag
  • Available in a wide range of colors, for color-coding and easy identification
  • Printable with all major thermal transfer barcode printers
  • Resistant to gamma irradiation (tested to up to 50 kGy)
General use paper labels for thermal-transfer printers are coated with a permanent adhesive and are available in a range of colours. They are ideal for use as shipping labels, colour-coding, price tagging, warehouses,More Product Information