Tisk…
Extended Exposure Xylene and Chemical-Resistant Thermal Transfer Labels
Dodavatel: GA
FTT-117C1-3WH FTT117NPC1-3WH FTT-125C1-4WH FTT-152C1-1WH FTT-152C3-3WH FTT-157C1-1WH FTT-159C1-1WH FTT188NPC3-3WH FTT-524C1-1WH FTT-545C1-1WH FTT-67C1-1WH FTT-98C1-1WH FTT-98C3-3WH
GAINFTT-117C1-3WHEA 2270 CZK
GAINFTT-117C1-3WH GAINFTT117NPC1-3WH GAINFTT-125C1-4WH GAINFTT-152C1-1WH GAINFTT-152C3-3WH GAINFTT-157C1-1WH GAINFTT-159C1-1WH GAINFTT188NPC3-3WH GAINFTT-524C1-1WH GAINFTT-545C1-1WH GAINFTT-67C1-1WH GAINFTT-98C1-1WH GAINFTT-98C3-3WH
Extended Exposure Xylene and Chemical-Resistant Thermal Transfer Labels
Štítky
Xylene and chemical-resistant labels for the identification of microscope slides that will undergo long-term chemical exposure and immersion.
  • Temperature range: −80 to +120 °C
  • Excellent stain resistance, ideal for microscope slides used in histology
  • Recommended for routine as well as special staining protocols
  • Compatible with automated slide processors
  • Waterproof and abrasion-resistant
  • Compatible with all common thermal transfer barcode printers
  • XAR-class resin ribbon recommended
Intended for use on flat surfaces, these solvent-resistant labels, alongMore Product Information