Tisk…
DYMO-compatible cryogenic labels
Dodavatel: GA
EG1F-072BE EG1F-072BL EG1F-072BR EG1F-072GA EG1F-072GR EG1F-072LE EG1F-072OR EG1F-072RE EG1F-072VI EG1F-072YE EG1F-081BE EG1F-081BL EG1F-081BR EG1F-081GA EG1F-081GR EG1F-081LE EG1F-081OR EG1F-081RE EG1F-081VI EG1F-081YE
GAINEG1F-072BEEA 2590 CZK
GAINEG1F-072BE GAINEG1F-072BL GAINEG1F-072BR GAINEG1F-072GA GAINEG1F-072GR GAINEG1F-072LE GAINEG1F-072OR GAINEG1F-072RE GAINEG1F-072VI GAINEG1F-072YE GAINEG1F-081BE GAINEG1F-081BL GAINEG1F-081BR GAINEG1F-081GA GAINEG1F-081GR GAINEG1F-081LE GAINEG1F-081OR GAINEG1F-081RE GAINEG1F-081VI GAINEG1F-081YE
DYMO-compatible cryogenic labels
Štítky
DYMO compatible, direct thermal cryo labels. For ultra-low and cryogenic temperature applications as low as –196 °C.
  • EG1F-081 series labels are resistant to gamma irradiation (tested to up to 50 kGy)
  • Ideal for the identification of 0,5 to 5 ml vials and tubes
  • Compatible with all DYMO label writers
  • Temperature range: –196 to 70 °C
  • Permanent adhesive
Ideal for identification and permanent storage of vials and containers in liquid nitrogen and laboratory freezers (–80, –50, –20 °C). Labels will also resist extreme cryogenic temperatures (–196 °C in aMore Product Information