Tisk…
Cryogenic labels for the identification of IVF straws
Dodavatel: GA
CST28NP.25C1WH CST28NPC3-1WH CST28NP.05C1WH CST28NP0.5C3WH CST29NPC1-1BL CST29NPC1-1GA CST29NPC1-1PI CST29NPC1-1RE CST29NPC1-1WH CST29NPC1-1YE CST29NPC3-1BL CST29NPC3-1GA CST29NPC3-1PI CST29NPC3-1RE CST29NPC3-1WH CST29NPC3-1YE CST29NP0.5C1PI CST29NP0.5C1RE CST29NP0.5C1WH CST29NP0.5C3PI CST29NP0.5C3RE CST29NP0.5C3WH CST79NPNC1-1WH CST79NPC3-1WH CST79NP0.5C1WH CST79NP0.5C3WH
GAINCST28NP.25C1WHEA 3280 CZK
GAINCST28NP.25C1WH GAINCST28NPC3-1WH GAINCST28NP.05C1WH GAINCST28NP0.5C3WH GAINCST29NPC1-1BL GAINCST29NPC1-1GA GAINCST29NPC1-1PI GAINCST29NPC1-1RE GAINCST29NPC1-1WH GAINCST29NPC1-1YE GAINCST29NPC3-1BL GAINCST29NPC3-1GA GAINCST29NPC3-1PI GAINCST29NPC3-1RE GAINCST29NPC3-1WH GAINCST29NPC3-1YE GAINCST29NP0.5C1PI GAINCST29NP0.5C1RE GAINCST29NP0.5C1WH GAINCST29NP0.5C3PI GAINCST29NP0.5C3RE GAINCST29NP0.5C3WH GAINCST79NPNC1-1WH GAINCST79NPC3-1WH GAINCST79NP0.5C1WH GAINCST79NP0.5C3WH
Cryogenic labels for the identification of IVF straws
Štítky
Printable with thermal-transfer barcode printers for secure sample identification and traceability.
  • Self-laminating, wrap-around labels provide enhanced resistance to abrasion, chemicals, scratching, and smudging
  • For short and long-term storage in vapor or liquid phase liquid nitrogen (–196 °C/–321 °F)
  • Specifically designed for the identification of cryo-preservation straws (0,25 ml and 0,5 ml) used in IVF laboratories and clinics
  • Available in standard white, as well as an assortment of colors for quick and easy identification via color-coding