Tisk…
Removable deep-freeze labels for microplates
Dodavatel: GA
AMA-11C1-2WH AMA-11C3-4WH AMA-223C1-2BL AMA-223C1-2GA AMA-223C1-2WH AMA-223C1-2YE AMA-223C3-4BL AMA-223C3-4GA AMA-223C3-4WH AMA-223C3-4YE
GAINAMA-11C1-2WHEA 2590 CZK
GAINAMA-11C1-2WH GAINAMA-11C3-4WH GAINAMA-223C1-2BL GAINAMA-223C1-2GA GAINAMA-223C1-2WH GAINAMA-223C1-2YE GAINAMA-223C3-4BL GAINAMA-223C3-4GA GAINAMA-223C3-4WH GAINAMA-223C3-4YE
Removable deep-freeze labels for microplates
Štítky
These micro-well plate labels are particularly useful for microbiology, genetics, and drug discovery laboratories.
  • Perfectly suited for the identification of microtiter plates
  • Able to withstand a wide temperature range: –80 to 72 °C (–112 to 161,6 °F)
  • Glove-friendly adhesive ensures labels can be removed without leaving any glue residue behind
Waterproof labels made of a durable thermoplastic film are ideal for use on cell culture plates and flasks, micro-well plates, as well as PCR plates and Petri dishes. These labels retain their shape when removedMore Product Information