Tisk…
Adhesive-free formalin resistant tags for thermal transfer printers, FormaliTAG™
Dodavatel: GA
ALA-25C1-125 ALA-25C3-500 ALA-64C1-125 ALA-64C3-500 ALA-76C1-125 ALA-76C3-500
GAINALA-25C1-125EA 1160 CZK
GAINALA-25C1-125 GAINALA-25C3-500 GAINALA-64C1-125 GAINALA-64C3-500 GAINALA-76C1-125 GAINALA-76C3-500
Adhesive-free formalin resistant tags for thermal transfer printers, FormaliTAG™
Štítky
Designed to be resistant to formalin, these tags can be used with formalin solutions (10 to 100%) and alcohols.
  • Will withstand direct exposure to harsh chemicals and solvents
  • Supplied in a continuous roll for ease of use and size selection
  • Able to withstand long-term immersion and storage in liquid preservatives
FormaliTAG™ labels are adhesive-free tags for thermal transfer printers that allow permanent and long-term identification of biological specimens and samples fixed and preserved in formaldehyde. Durable and reliable, they are perfect forMore Product Information