Tisk…
Cryogenic labels for frozen vials and containers, CryoSTUCK®
Dodavatel: GA
AEA-1C1-1WH AEA-2NOTC1-1WH AEA-2NOTC3-1WH AEA-3C1-1WH AEA-3C3-1WH AEA-4C1-1WH AEA-4C3-1WH AEA-5NOTC1-1WH AEA-5NOTC3-1WH AEA-6C1-1WH AEA-6C3-1WH AEA-7C1-1WH AEA-7C3-2WH
GAINAEA-1C1-1WHEA 3910 CZK
GAINAEA-1C1-1WH GAINAEA-2NOTC1-1WH GAINAEA-2NOTC3-1WH GAINAEA-3C1-1WH GAINAEA-3C3-1WH GAINAEA-4C1-1WH GAINAEA-4C3-1WH GAINAEA-5NOTC1-1WH GAINAEA-5NOTC3-1WH GAINAEA-6C1-1WH GAINAEA-6C3-1WH GAINAEA-7C1-1WH GAINAEA-7C3-2WH
Cryogenic labels for frozen vials and containers, CryoSTUCK®
Štítky
Cryogenic labels, with a clear overlap area, designed to be applied directly, and rapidly adhere to frozen cryo vials, tubes, cryo and freezer boxes, as well as other containers which are difficult to label under deep-freeze conditions (–80 °C/–112 °F).
  • Adhere quickly to already frozen surfaces (no thawing required)
  • Ideal for long-term storage in cryogenic conditions (–196 °C)
  • Available in clear, white and cover-up/blockout versions
  • Extra-permanent adhesive, ensures strong adhesion even to wet surfaces
Designed for long-term preservation in liquidMore Product Information