Tisk…
Permanent Cryogenic Labels for Laser Printers
Dodavatel: GA
A4CL-111T1-WH A4CL-112T1-WH A4CL-11T1-WH A4CL-12T1-WH A4CL-23T1-A2 A4CL-23T1-BL A4CL-23T1-GA A4CL-23T1-OR A4CL-23T1-PI A4CL-23T1-RE A4CL-23T1-SI A4CL-23T1-WH A4CL-23T1-YE A4CL-28T1-WH A4CL-32T1-WH A4CL-36T1-WH A4CL-3T1-WH A4CL-48T1-A2 A4CL-48T1-BL A4CL-48T1-GA A4CL-48T1-OR A4CL-48T1-PI A4CL-48T1-RE A4CL-48T1-SI A4CL-48T1-WH A4CL-48T1-YE A4CL-6T1-WH A4CL-72T1-WH A4CL-74T1-BE A4CL-74T1-BL A4CL-74T1-BR A4CL-74T1-GA A4CL-74T1-LA A4CL-74T1-MT A4CL-74T1-OR A4CL-74T1-PI A4CL-74T1-RE A4CL-74T1-SI A4CL-74T1-VI A4CL-74T1-WH A4CL-74T1-YE A4CL-81T1-WH A4CL-82T1-WH A4CL-83T1-WH A4CL-84T1-WH A4CL-85T1-WH A4CL-8T1-WH A4CL-97T1-WH A4CL-9T1-WH
GAINA4CL-111T1-WHEA 2020 CZK
GAINA4CL-111T1-WH GAINA4CL-112T1-WH GAINA4CL-11T1-WH GAINA4CL-12T1-WH GAINA4CL-23T1-A2 GAINA4CL-23T1-BL GAINA4CL-23T1-GA GAINA4CL-23T1-OR GAINA4CL-23T1-PI GAINA4CL-23T1-RE GAINA4CL-23T1-SI GAINA4CL-23T1-WH GAINA4CL-23T1-YE GAINA4CL-28T1-WH GAINA4CL-32T1-WH GAINA4CL-36T1-WH GAINA4CL-3T1-WH GAINA4CL-48T1-A2 GAINA4CL-48T1-BL GAINA4CL-48T1-GA GAINA4CL-48T1-OR GAINA4CL-48T1-PI GAINA4CL-48T1-RE GAINA4CL-48T1-SI GAINA4CL-48T1-WH GAINA4CL-48T1-YE GAINA4CL-6T1-WH GAINA4CL-72T1-WH GAINA4CL-74T1-BE GAINA4CL-74T1-BL GAINA4CL-74T1-BR GAINA4CL-74T1-GA GAINA4CL-74T1-LA GAINA4CL-74T1-MT GAINA4CL-74T1-OR GAINA4CL-74T1-PI GAINA4CL-74T1-RE GAINA4CL-74T1-SI GAINA4CL-74T1-VI GAINA4CL-74T1-WH GAINA4CL-74T1-YE GAINA4CL-81T1-WH GAINA4CL-82T1-WH GAINA4CL-83T1-WH GAINA4CL-84T1-WH GAINA4CL-85T1-WH GAINA4CL-8T1-WH GAINA4CL-97T1-WH GAINA4CL-9T1-WH
Permanent Cryogenic Labels for Laser Printers
Štítky
Permanent cryogenic laser labels for long-term storage in liquid phase and vapor phase liquid nitrogen (−196 °C), lab freezers (−80 °C), and on dry ice. Supplied in A4 sheet format for printing in standard desktop laser printers, these cryogenic labels do not jam printers and withstand UV and gamma irradiation.
  • Temperature range: −196 to 120 °C
  • Permanent adhesion, especially on curved surfaces (vials and microtubes)
  • Withstands extreme cryogenic temperatures (liquid phase and vapor phase liquid nitrogen)
  • Printable by all major desktop laser printers