Tisk…
KingFisher™ Apex purification system
Dodavatel: Thermo Fisher Scientific
97002560 97002570 97002580 5400910 5400930 5400920 5400940
FINN97002560EA 10050 CZK
FINN97002560 FINN97002570 FINN97002580 731-0708 731-0710 731-0709 731-0711
KingFisher™ Apex purification system
Atomatizované systémy k purifikaci nukleových kyselin
The Thermo Scientific KingFisher Apex represents the pinnacle of automated purification for DNA, RNA, proteins, and cells. Building on decades of KingFisher product value and excellence, we’ve combined superior instrument capabilities with complete touch screen-based control to deliver unrivalled flexibility and performance. The KingFisher Apex makes sample prep simple.
  • Touch screen control: A modern touch screen display and an intuitive interface puts you in control of instrument operation
  • Choice of plate or tube formats: Wide 20 µl to 5 ml volume range
More Product Information