Tisk…
ECOSAN® Neutral Skin Wash
Dodavatel: FAWECO HAUTSCHUTZ
13897-015 13897-004 13897-003 13897-024
FAWE13897-015EA 0 CZK
FAWE13897-015 FAWE13897-004 FAWE13897-003 FAWE13897-024
ECOSAN® Neutral Skin Wash
Péče o pokožku Mýdla a čističe pokožky
Neutral cleansing syndet, also suitable for the food industry.
  • Transparent skin wash
  • Colorant free
  • Soap and alkali free
  • Skin-neutral pH value
  • Perfume and fragrance free
BLUE ANGEL environmental label and EU Ecolabel for 1000 ml Neptune bottles and 2000 ml collapsible bottles.
Certifikace: HACCP compliant.